Please login to your online account to display your discounted pricing

Sodium ethoxide, pure, 21% in ethanol, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC213221000

 View more versions of this product

Catalog No. AC213221000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 68.04
Quantity 100g
Color Yellow to brown
Merck Index 15, 8749
Beilstein 01, 311
Assay 20.5 to 21.5% (NaOC2H5)
TSCA TSCA
EINECS Number 205-487-5
Purity Grade Pure
Density 0.8680g/mL
Fieser 01,1065; 03,265; 04,451; 06,540
Sodium Hydroxide and Sodium Carbonate 0.5% max.
Storage Information May darken on storage with no change in quality
Alkalinity 20.5 to 22.0 (as NaOC2H5)
CAS 64-17-5
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Reacts violently with water.
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material Sodium ethoxide
Linear Formula C2H5ONa
Molecular Formula C2H5NaO
Name Note 21% in Ethanol
MDL Number MFCD00012417
CAS 141-52-6

Specifications

Chemical Name or Material Sodium ethoxide
Name Note 21% in Ethanol
CAS 141-52-6
CAS 64-17-5
Purity Grade Pure
Assay 20.5 to 21.5% (NaOC2H5)
Molecular Formula C2H5NaO
Linear Formula C2H5ONa
Formula Weight 68.04
MDL Number MFCD00012417
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Reacts violently with water.

SDS & Safety Documents