missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium ethoxide, 21% w/w soln. in ethanol, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer:  Alfa Aesar™ 45590AE

 View more versions of this product

Catalog No. AA45590AE



Description

Description

  • Packaged under Argon
Specifications

Specifications

Sodium ethoxide
20°C (68°F)
1.3850
100mL
C2H5NaO
MFCD00012417
3593646
sodium ethoxide, sodium ethylate, sodium ethanolate, sodiumethoxide, ethoxysodium, ethanol, sodium salt, caustic alcohol, naoet, etona, ethanol, sodium salt 1:1
QDRKDTQENPPHOJ-UHFFFAOYSA-N
sodium;ethanolate
2723922
68.05
0.868g/mL
ChemSeal™ bottles
Moisture Sensitive
141-52-6
NaOCH2CH3
UN3274
14,8613
21% w/w soln. in ethanol
CC[O-].[Na+]
68.051
CHEBI:52096
Liquid

Chemical Identifiers

141-52-6
68.051
QDRKDTQENPPHOJ-UHFFFAOYSA-N
2723922
sodium;ethanolate
C2H5NaO
MFCD00012417
sodium ethoxide, sodium ethylate, sodium ethanolate, sodiumethoxide, ethoxysodium, ethanol, sodium salt, caustic alcohol, naoet, etona, ethanol, sodium salt 1:1
CHEBI:52096
CC[O-].[Na+]
Safety and Handling

Safety and Handling

GHS H Statement
H224-H314-H318
Extremely flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLATES SOLUTION, N.O.S.

TSCA : TSCA

storageNote1 :

  • Packaged under Argon

SDS Documents