missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium ethoxide, 21% in ethanol, AcroSeal™, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 42930100

 View more versions of this product

Catalog No. AC429301000


 

Specifications Description & Specifications

Specifications

Sodium ethoxide
141-52-6,64-17-5
C2H5ONa
MFCD00012417
AcroSeal™ Glass Bottle
21% in ethanol
C2H5NaO
68.04
0.868g/mL
100mL

Chemical Identifiers

141-52-6,64-17-5
C2H5ONa
MFCD00012417
C2H5NaO
68.04
Sodium ethoxide, 21% in ethanol

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger