missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Dodecylsulfate, 10% Solution, Ultrapure, Thermo Scientific Chemicals
Supplier: Thermo Fisher Scientific J77504AE
Specifications
Sodium Dodecylsulfate | |
Solution | |
C12H25NaO4S | |
100 mL | |
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O | |
288.38 | |
CHEBI:8984 |
151-21-3 | |
10% Solution | |
MFCD00036175 | |
DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
sodium dodecyl sulfate | |
3423265 | |
Nalgene bottle |
Chemical Identifiers
151-21-3 | |
288.38 | |
DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
CHEBI:8984 |
C12H25NaO4S | |
MFCD00036175 | |
3423265 | |
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
Safety and Handling
P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362
H315-H318
EINECSNumber : 205-788-1
RTECSNumber : WT1050000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only