Please login to your online account to display your discounted pricing

Sodium Dodecyl Sulfate, Ultrapure Bioreagent, J.T.Baker™

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

$117.79 - $1,027.84

Chemical Identifiers

Chemical Name or Material Sodium Dodecyl Sulfate
CAS 151-21-3
Purity Grade Ultrapure Bioreagent
Molecular Formula CH3(CH2)11OSO3Na
Formula Weight 288.38
Synonym SDS
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004080

 
avantor performance
409504
Poly Bottle 100g Each for $117.79

02004082

 
avantor performance
409502
Poly bottle 1kg Each for $576.30

02004084

 
avantor performance
409505
Poly Pail 2kg Each for $1,027.84
Description & Specifications

Specifications

Formula Weight 288.38
Chemical Name or Material Sodium Dodecyl Sulfate
CAS 151-21-3
Purity Grade Ultrapure Bioreagent
Molecular Formula CH3(CH2)11OSO3Na
Packaging Poly Bottle
Synonym SDS
Quantity 100g
Flash Point 170.00°C
Specific Gravity 1L = 0.40kg

    For Electrophoresis and Protein Solubilization; For Laboratory,Research,or Manufacturing Use