Please login to your online account to display your discounted pricing

Sodium Dodecyl Sulfate, Ultrapure Bioreagent, J.T.Baker™


High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

Quantity 100g
Packaging Poly Bottle

Manufacturer: avantor performance  409504

 View more versions of this product

Catalog No. 02004080

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Sodium Dodecyl Sulfate
CAS 151-21-3
Purity Grade Ultrapure Bioreagent
Molecular Formula CH3(CH2)11OSO3Na
Formula Weight 288.38
Packaging Poly Bottle
Synonym SDS
Flash Point 170.00°C
Quantity 100g
Specific Gravity 1L = 0.40kg

Specifications

Chemical Name or Material Sodium Dodecyl Sulfate
CAS 151-21-3
Purity Grade Ultrapure Bioreagent
Molecular Formula CH3(CH2)11OSO3Na
Formula Weight 288.38
Synonym SDS

    For Electrophoresis and Protein Solubilization