Please login to your online account to display your discounted pricing

Sodium Dodecyl Sulfate (SDS), Micropellets, Fisher BioReagents


Sodium Dodecyl Sulfate is the most commonly used detergent in protein purification and electrophoresis

Quantity 10g
Packaging Poly Bottle

Manufacturer: thermo fisher scientific  BP820010

 View more versions of this product

Catalog No. BP820010

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Molecular Biology Grade
Melting Point 204°C
Quantity 10g
Phosphate ≤0.0005%
Purity ≥99.0%
Chloride ≤0.1%
Packaging Poly Bottle
Assay ≥98.0% as C12 alcohol
Color Off-white
Iron (Fe) ≤0.0001%
Appearance White micro-pellets
DNase DNase free
Chemical Name or Material Sodium Dodecyl Sulfate (SDS), Micropellets
Protease Protease free
Identification Pass test
CAS 151-21-3
Copper (Cu) ≤0.0005%
Heavy Metals (as Pb) ≤0.0005%
Molecular Formula C12H25NaO4S
Physical Form Solid
Electrophoresis Pass test
Absorbance ≤0.10 (0.1% Solution) at 260nm
Sulfate <0.1%
pH 8.5 to 10
Assay Percent Range >99%
Recommended Storage Store at room temperature

Specifications

Chemical Name or Material Sodium Dodecyl Sulfate (SDS), Micropellets
CAS 151-21-3
Purity Grade Molecular Biology Grade
Physical Form Solid
Assay ≥98.0% as C12 alcohol
Assay Percent Range >99%
Accessories

Store at room temperature

SDS & Safety Documents