missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
SDS Solution, 20% Sodium Dodecyl Sulfate Solution, Molecular Biology/Electrophoresis, Fisher BioReagents™
Supplier: Fisher BioReagents V35202
Description
SDS Solution, 20% Sodium Dodecyl Sulfate Solution, Molecular Biology/Electrophoresis
- DNase, RNase, and protease-free premixed solution containing 20% SDS for protein extraction and DNA extraction procedures
- Reduce preparation time by using this premade SDS solution
- SDS is a commonly used detergent in protein purification, polyacrylamide gel electrophoresis (PAGE), and other molecular biology applications
- SDS is also used to disrupt cell membranes and dissociate nucleic acid-protein complexes in DNA extraction procedures
- Intended for research use only; not for human or animal diagnostic purposes
Specifications
0.4 max. (0.1% solution) at 230nm, 0.1 max. (0.1% solution) at 280nm | |
Colorless | |
151-21-3,7732-18-5 | |
100°C | |
1.01g/cm³ | |
19.5 to 20.5% | |
CH3(CH2)10CH2OSO3Na | |
200 mL | |
Sodium lauryl sulfate, SDS | |
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O | |
288.38 | |
3423265 | |
288.38 | |
Electrophoresis/Molecular Biology |
Sodium Dodecyl Sulfate | |
20% Solution | |
Liquid | |
DNase-, RNase- and Protease-Free | |
(as Fatty Alcohol Sulfate): ≥99% | |
C12H25NaO4S | |
MFCD00036175 | |
15, 8768 | |
DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
sodium;dodecyl sulfate | |
Pass Test | |
CHEBI:8984 | |
≥99% | |
Poly Bottle |
Chemical Identifiers
151-21-3 | |
288.38 | |
DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
3423265 | |
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
C12H25NaO4S | |
MFCD00036175 | |
Sodium lauryl sulfate, SDS | |
CHEBI:8984 |
Safety and Handling
Recommended Storage : RT