Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium dodecatungstophosphate hydrate

$54.69 - $566.54

Chemical Identifiers

Chemical Name or Material Sodium dodecatungstophosphate hydrate
CAS 51312-42-6
Molecular Formula Na3PO4·12WO3·xH2O
Formula Weight 2946.12 (Anhydrous)
MDL Number MFCD00151658
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1157514

  SDS
alfa aesar™
A1157514
25g Each for $54.69

AAA1157536

  SDS
alfa aesar™
A1157536
500g Each for $566.54
Description & Specifications

Specifications

Formula Weight 2946.12 (Anhydrous)
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number TH5775000
Chemical Name or Material Sodium dodecatungstophosphate hydrate
Health Hazard 1 Danger
TSCA TSCA
Quantity 25g
Molecular Formula Na3PO4·12WO3·xH2O
MDL Number MFCD00151658
Melting Point 350°C
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 51312-42-6
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.