Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium dodecamolybdophosphate hydrate, 90+%

$48.34 - $514.12

Chemical Identifiers

Chemical Name or Material Sodium dodecamolybdophosphate hydrate
CAS 1313-30-0
Molecular Formula Na3PO4·12MoO3·xH2O
Formula Weight 1891.22 (Anhydrous)
MDL Number MFCD00150420
Merck Index 14,8664
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1155114

 
alfa aesar™
A1155114
25g Each for $48.34

AAA1155136

 
alfa aesar™
A1155136
500g Each for $514.12
Description & Specifications

Specifications

Formula Weight 1891.22 (Anhydrous)
Chemical Name or Material Sodium dodecamolybdophosphate hydrate
Assay ≥90%
Molecular Formula Na3PO4·12MoO3·xH2O
MDL Number MFCD00150420
CAS 1313-30-0
Merck Index 14,8664
Quantity 25g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number WD6600000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.