Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium diphenylamine-4-sulfonate, 0.005M solution in water

Redox indicator

$36.03 - $97.12

Chemical Identifiers

Chemical Name or Material Sodium diphenylamine-4-sulfonate
CAS 6152-67-6
Molecular Formula C12H10NNaO3S
Formula Weight 271.27
MDL Number MFCD00036468
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42809AE

  SDS
alfa aesar™
42809AE
100mL Each for $36.03

AA42809AP

  SDS
alfa aesar™
42809AP
500mL Each for $97.12
Description & Specifications

Specifications

Formula Weight 271.27
Chemical Name or Material Sodium diphenylamine-4-sulfonate
TSCA TSCA
Physical Form Liquid
Molecular Formula C12H10NNaO3S
MDL Number MFCD00036468
CAS 6152-67-6
Concentration 0.005M solution in water
Quantity 100mL
Odor Odorless