missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium dihydrogen phosphate monohydrate, 99.998% (metals basis), Alfa Aesar™ Puratronic™

Manufacturer:  Thermo Fisher Scientific 1087022

 View more versions of this product

Catalog No. AA1087022



Specifications

Specifications

Sodium dihydrogen phosphate monohydrate
99.998% (metals basis)
MFCD00149208
monosodium phosphate monohydrate
O.OP(=O)(O)[O-].[Na+]
137.991
137.99 (119.98 Anhydrous)
Odorless
100g
10049-21-5
H4NaO5P
White powder
BBMHARZCALWXSL-UHFFFAOYSA-M
sodium;dihydrogen phosphate;hydrate
516949
Powder
White

Chemical Identifiers

10049-21-5
137.991
BBMHARZCALWXSL-UHFFFAOYSA-M
516949
O.OP(=O)(O)[O-].[Na+]
H4NaO5P
MFCD00149208
monosodium phosphate monohydrate
sodium;dihydrogen phosphate;hydrate
Safety and Handling

Safety and Handling

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

EINECSNumber : 231-449-2

Documents