Please login to your online account to display your discounted pricing

Sodium Diethyldithiocarbamate, Trihydrate, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$265.97 - $642.24

Chemical Identifiers

Chemical Name or Material Sodium Diethyldithiocarbamate
CAS 20624-25-3
Name Note Trihydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula (C2H5)2NCSSNa·3H2O
Formula Weight 225.31
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004277

 
avantor performance
862404
Poly Bottle 125g Each for $265.97

02004278

 
avantor performance
862401
Poly Bottle 500g Each for $642.24
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 225.31
Name Note Trihydrate
Packaging Poly Bottle
Specific Gravity 1L = 1.10kg
Molecular Formula (C2H5)2NCSSNa·3H2O
CAS 20624-25-3
Quantity 125g
Chemical Name or Material Sodium Diethyldithiocarbamate