Please login to your online account to display your discounted pricing

Sodium Diethyldithiocarbamate, Certified, 0.1% (w/v), LabChem™

$77.97 - $77.97

Chemical Identifiers

Chemical Name or Material Sodium Diethyldithiocarbamate
CAS 7732-18-5
CAS Max % 99.9
CAS 20624-25-3
CAS Max % 0.1
Purity Grade Certified Grade
Linear Formula C5H10NNaS2·3H2O
Formula Weight 225.3
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC237201

 
lab chem
LC237201
500mL Each for $77.97
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 225.3
Chemical Name or Material Sodium Diethyldithiocarbamate
Identification Passes Test
Packaging Amber Poly
Physical Form Liquid
Assay 0.1% (w/v)
Solubility Soluble in water
Specific Gravity 1g/mL
Storage Information Room Temperature
Linear Formula C5H10NNaS2·3H2O
Suitability Passes Test
CAS 7732-18-5
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide; Sulfur compounds
Color Clear, colorless
Quantity 500mL
Health Hazard 1 Warning
CAS Max % 99.9
Health Hazard 2 GHS H Statement
May cause an allergic skin reaction.
CAS 20624-25-3
Health Hazard 3 GHS P Statement
Avoid breathing mist, vapors, spray.
Wear protective gloves, eye protection.
Contaminated work clothing must not be allowed out of the workplace.
If on skin: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
Take off contaminated clothing and wash it before reuse.
Dispose of contents/container to comply with local, state and federal regulations.
CAS Max % 0.1
Density 1g/mL
Safety and Handling

Warning

GHS P Statement
Avoid breathing mist, vapors, spray.
Wear protective gloves, eye protection.
Contaminated work clothing must not be allowed out of the workplace.
If on skin: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
Take off contaminated clothing and wash it before reuse.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
May cause an allergic skin reaction.