Please login to your online account to display your discounted pricing

Sodium Diethyldithiocarbamate, 0.1% (w/v) Aqueous Solution, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Sodium Diethyldithiocarbamate
Name Note 0.1% (w/v) Aqueous Solution
CAS 20624-25-3
CAS 7732-18-5
Purity Grade ASTM/ACS Certified Reagent
Physical Form Liquid
Assay Percent Range 0.1% (w/v)
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R72406001A

 
ricca chemical company
RIR72406001A
Poly natural 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R724060020F

 
ricca chemical company
RIR724060020F
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R72406004A

 
ricca chemical company
RIR72406004A
Poly natural 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Sodium Diethyldithiocarbamate
Name Note 0.1% (w/v) Aqueous Solution
CAS 20624-25-3
CAS 7732-18-5
Purity Grade ASTM/ACS Certified Reagent
Physical Form Liquid
Assay Percent Range 0.1% (w/v)
Packaging Poly natural
Solubility Solubility in Water: Infinite
Solvent or Matrix Aqueous Solution
Color Clear colorless
Quantity 1L