missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Diethyldithiocarbamate, 0.1% (w/v) Aqueous Solution, Ricca Chemical

Manufacturer:  Ricca Chemical Company RIR72406004A

 View more versions of this product

Catalog No. R72406004A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Sodium Diethyldithiocarbamate
20624-25-3,7732-18-5
C5H16NNaO3S2
WWGXHTXOZKVJDN-UHFFFAOYSA-M
sodium;N,N-diethylcarbamodithioate;trihydrate
517546
ASTM/ACS Certified Reagent
Solubility in Water: Infinite
Colorless
0.1% (w/v) Aqueous Solution
0.1% (w/v)
sodium diethyldithiocarbamate trihydrate, dithiocarb trihydrate, cupral, diethyldithiocarbamic acid sodium salt trihydrate, ditiocarb sodium trihydrate, unii-04574exf2c, diethyldithiocarbamate sodium trihydrate, carbamic acid, diethyldithio-, sodium salt, trihydrate, diethyldithiocarbamate sodium salt trihydrate, diethyldithiokarbaman sodny trihydrat czech
CCN(CC)C(=S)[S-].O.O.O.[Na+]
225.297
Liquid
Poly natural
Aqueous Solution
4L

Chemical Identifiers

20624-25-3
225.297
sodium diethyldithiocarbamate trihydrate, dithiocarb trihydrate, cupral, diethyldithiocarbamic acid sodium salt trihydrate, ditiocarb sodium trihydrate, unii-04574exf2c, diethyldithiocarbamate sodium trihydrate, carbamic acid, diethyldithio-, sodium salt, trihydrate, diethyldithiocarbamate sodium salt trihydrate, diethyldithiokarbaman sodny trihydrat czech
sodium;N,N-diethylcarbamodithioate;trihydrate
C5H16NNaO3S2
WWGXHTXOZKVJDN-UHFFFAOYSA-M
517546
CCN(CC)C(=S)[S-].O.O.O.[Na+]
Documents