Please login to your online account to display your discounted pricing

Sodium Diethyldithiocarbamate, 0.1% (w/v) Aqueous Solution, Ricca Chemical


Quantity 20L
Packaging Cubitainer

Manufacturer: ricca chemical company  RIR724060020F

 View more versions of this product

Catalog No. R724060020F

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Sodium Diethyldithiocarbamate
Name Note 0.1% (w/v) Aqueous Solution
CAS 20624-25-3
CAS 7732-18-5
Purity Grade ASTM/ACS Certified Reagent
Physical Form Liquid
Assay Percent Range 0.1% (w/v)
Packaging Cubitainer
Solubility Solubility in Water: Infinite
Solvent or Matrix Aqueous Solution
Color Clear colorless
Quantity 20L

Specifications

Chemical Name or Material Sodium Diethyldithiocarbamate
Name Note 0.1% (w/v) Aqueous Solution
CAS 20624-25-3
CAS 7732-18-5
Purity Grade ASTM/ACS Certified Reagent
Physical Form Liquid
Assay Percent Range 0.1% (w/v)