Learn More
Sodium dichromate dihydrate, Reagent Grade, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 012611A3
Description
Sodium dichromate is used as a strong oxidizing agent in organic synthesis. It is used in metal finishing as an aid of corrosion resistance, manufacture of inorganic chromate pigments and in the preparation of colored glass and ceramic glazes. It is used as a mordant for acidic dyes to improve their color-fast properties and as a corrosion inhibitors in undercoats and primers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium dichromate dihydrate | |
7789-12-0 | |
Crystalline | |
Cr2H4Na2O9 | |
sodium dichromate dihydrate, unii-52125xyy2a, ccris 6344, disodium chromate dihydrate, sodium dichromate vi dihydrate, sodium dichromate dihydrate vi, chromic acid h2cr2o7 , disodium salt, dihydrate, dichromic acid h2cr2o7 , disodium salt, dihydrate, dsstox_cid_12061, dsstox_rid_78903 | |
JYDRNIYTFCBIFC-UHFFFAOYSA-N | |
disodium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium;dihydrate | |
108005 | |
Reagent | |
Red crystals |
100°C-2H2O | |
Red | |
2 kg | |
MFCD00149166 | |
Soluble in waterSoluble in water. Insoluble in alcohol. | |
O.O.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[Na+].[Na+] | |
297.995 | |
298.00 (261.97 Anhydrous) | |
Hygroscopic | |
2.348 |
Chemical Identifiers
7789-12-0 | |
297.995 | |
JYDRNIYTFCBIFC-UHFFFAOYSA-N | |
108005 | |
O.O.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[Na+].[Na+] |
Cr2H4Na2O9 | |
MFCD00149166 | |
sodium dichromate dihydrate, unii-52125xyy2a, ccris 6344, disodium chromate dihydrate, sodium dichromate vi dihydrate, sodium dichromate dihydrate vi, chromic acid h2cr2o7 , disodium salt, dihydrate, dichromic acid h2cr2o7 , disodium salt, dihydrate, dsstox_cid_12061, dsstox_rid_78903 | |
disodium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium;dihydrate |
Safety and Handling
P201-P202-P210-P220-P221-P260-P264b-P270-P271-P272-P280g-P281-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
H272-H301-H312-H314-H317-H330-H334-H335-H340-H350-H360FD-H372
DOTInformation : DOT Class: 5.1, Packaging Group: II
EINECSNumber : 234-190-3
RTECSNumber : HX7750000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only