Please login to your online account to display your discounted pricing

Sodium Dichromate Dihydrate (Granular/Technical), Fisher Chemical


Quantity 3kg
Packaging Poly Bottle

Manufacturer: fisher scientific company  S2343

 View more versions of this product

Catalog No. S234-3

  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Technical Grade
Formula Weight 298
Boiling Point 400°C
Melting Point 357°C
Quantity 3kg
DOT Information DOT Class 5.1, : Oxidizer
Physical Form Solid
Molecular Formula Cr2Na2O7 · 2 H2O
Viscosity [NULL]
Color Orange
pH 3.5 to 3.9
Identification Pass Test
CAS 7789-12-0
Health Hazard 1 Signal Word:DANGER!
MDL Number MFCD00149166
Chemical Name or Material Sodium Dichromate Dihydrate
Packaging Poly Bottle
ChemAlert Storage Symbol Yellow
Health Hazard 3 Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Cancer hazard. Causes burns by all exposure routes. Very toxic by inhalation. Toxic if swallowed. Harmful in contact with skin.

Specifications

Chemical Name or Material Sodium Dichromate Dihydrate
CAS 7789-12-0
Purity Grade Technical Grade
Physical Form Solid
Molecular Formula Cr2Na2O7 · 2 H2O
Formula Weight 298
MDL Number MFCD00149166
Accessories
 

Signal Word:DANGER!

Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Cancer hazard. Causes burns by all exposure routes. Very toxic by inhalation. Toxic if swallowed. Harmful in contact with skin.

SDS & Safety Documents