Please login to your online account to display your discounted pricing

Sodium Dichromate Dihydrate (Granular/Technical), Fisher Chemical


Quantity 10kg
Packaging Poly Pail

Manufacturer: fisher scientific company  S23410

 View more versions of this product

Catalog No. S234-10

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Technical Grade
Formula Weight 298
Boiling Point 400°C
Melting Point 357°C
Quantity 10kg
DOT Information DOT Class 5.1, : Oxidizer
Physical Form Solid
Molecular Formula Cr2Na2O7 · 2 H2O
Viscosity [NULL]
Color Orange
pH 3.5 to 3.9
Packaging Poly Pail
Chemical Name or Material Sodium Dichromate Dihydrate
Identification Pass Test
MDL Number MFCD00149166
CAS 7789-12-0
ChemAlert Storage Symbol Yellow
Health Hazard 1 Signal Word:DANGER!
Health Hazard 3 Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Cancer hazard. Causes burns by all exposure routes. Very toxic by inhalation. Toxic if swallowed. Harmful in contact with skin.

Specifications

Chemical Name or Material Sodium Dichromate Dihydrate
CAS 7789-12-0
Purity Grade Technical Grade
Physical Form Solid
Molecular Formula Cr2Na2O7 · 2 H2O
Formula Weight 298
MDL Number MFCD00149166
Accessories
 

Signal Word:DANGER!

Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Cancer hazard. Causes burns by all exposure routes. Very toxic by inhalation. Toxic if swallowed. Harmful in contact with skin.

SDS & Safety Documents