Please login to your online account to display your discounted pricing

Sodium Dichromate Dihydrate (Granular/Technical), Fisher Chemical

$211.71 - $5,169.57

Chemical Identifiers

Chemical Name or Material Sodium Dichromate Dihydrate
CAS 7789-12-0
Purity Grade Technical Grade
Physical Form Solid
Molecular Formula Cr2Na2O7 · 2 H2O
Formula Weight 298
MDL Number MFCD00149166
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

S234-10

  SDS
fisher scientific company
S23410
Poly Pail 10kg Each for $1,882.07

S234-3

  SDS
fisher scientific company
S2343
Poly Bottle 3kg Case of 4 Each for $5,169.57

S234-500

  SDS
fisher scientific company
S234500
Poly Bottle 500g Each for $211.71
Case of 6 Each for $1,270.26
Description & Specifications

Specifications

Purity Grade Technical Grade
Formula Weight 298
Boiling Point 400°C
Melting Point 357°C
Quantity 10kg
DOT Information DOT Class 5.1, : Oxidizer
Physical Form Solid
Molecular Formula Cr2Na2O7 · 2 H2O
Viscosity [NULL]
Color Orange
pH 3.5 to 3.9
Packaging Poly Pail
Chemical Name or Material Sodium Dichromate Dihydrate
Identification Pass Test
MDL Number MFCD00149166
CAS 7789-12-0
ChemAlert Storage Symbol Yellow
Health Hazard 1 Signal Word:DANGER!
Health Hazard 3 Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Cancer hazard. Causes burns by all exposure routes. Very toxic by inhalation. Toxic if swallowed. Harmful in contact with skin.
Safety and Handling
 

Signal Word:DANGER!

Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Cancer hazard. Causes burns by all exposure routes. Very toxic by inhalation. Toxic if swallowed. Harmful in contact with skin.