Please login to your online account to display your discounted pricing

Sodium Dichromate, Dihydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 125g
Packaging Poly Bottle

Manufacturer: avantor performance  367204

 View more versions of this product

Catalog No. 02004078

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystals
Formula Weight 298
Purity Grade Baker Analyzed™ ACS Reagent
Chemical Name or Material Sodium Dichromate
CAS 7789-12-0
Molecular Formula Na2Cr2O7·2H2O
Name Note Dihydrate
Specific Gravity 1L = 2.35kg
Quantity 125g
Packaging Poly Bottle

Specifications

Chemical Name or Material Sodium Dichromate
CAS 7789-12-0
Name Note Dihydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula Na2Cr2O7·2H2O
Formula Weight 298