Please login to your online account to display your discounted pricing

Sodium Dichromate, Dihydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$259.91 - $1,626.16

Chemical Identifiers

Chemical Name or Material Sodium Dichromate
CAS 7789-12-0
Name Note Dihydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula Na2Cr2O7·2H2O
Formula Weight 298
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004076

 
avantor performance
367201
Poly Bottle 500g Each for $491.21

02004077

 
avantor performance
367205
Poly Bottle 2.5kg Each for $1,626.16

02004078

 
avantor performance
367204
Poly Bottle 125g Each for $259.91
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Physical Form Crystals
Formula Weight 298
Chemical Name or Material Sodium Dichromate
CAS 7789-12-0
Molecular Formula Na2Cr2O7·2H2O
Name Note Dihydrate
Packaging Poly Bottle
Specific Gravity 1L = 2.35kg
Quantity 500g