Learn More
Sodium dichromate dihydrate, ACS reagent, 99.5-100.5%, Thermo Scientific Chemicals
$77.39 - $680.46
Chemical Identifiers
CAS | 7789-12-0 |
---|---|
Molecular Formula | Cr2Na2O7·2H2O |
Molecular Weight (g/mol) | 298 |
InChI Key | JYDRNIYTFCBIFC-UHFFFAOYSA-N |
Synonym | sodium dichromate dihydrate, unii-52125xyy2a, ccris 6344, disodium chromate dihydrate, sodium dichromate vi dihydrate, sodium dichromate dihydrate vi, chromic acid h2cr2o7 , disodium salt, dihydrate, dichromic acid h2cr2o7 , disodium salt, dihydrate, dsstox_cid_12061, dsstox_rid_78903 |
PubChem CID | 108005 |
IUPAC Name | disodium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium;dihydrate |
SMILES | O.O.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[Na+].[Na+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC385631000
|
Thermo Scientific Chemicals
385631000 |
100 g | Plastic bottle |
Each for $77.39
|
|
||||
AC385635000
|
Thermo Scientific Chemicals
385635000 |
500 g | Plastic bottle |
Each for $201.01
|
|
||||
AC385630025
|
Thermo Scientific Chemicals
385630025 |
2.5 kg | Plastic bottle |
Each for $680.46
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7789-12-0 | |
298 | |
sodium dichromate dihydrate, unii-52125xyy2a, ccris 6344, disodium chromate dihydrate, sodium dichromate vi dihydrate, sodium dichromate dihydrate vi, chromic acid h2cr2o7 , disodium salt, dihydrate, dichromic acid h2cr2o7 , disodium salt, dihydrate, dsstox_cid_12061, dsstox_rid_78903 | |
disodium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium;dihydrate |
Cr2Na2O7·2H2O | |
JYDRNIYTFCBIFC-UHFFFAOYSA-N | |
108005 | |
O.O.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[Na+].[Na+] |
Specifications
400.0°C | |
7789-12-0 | |
99.5-100.5% | |
Na2Cr2O7·2H2O | |
sodium dichromate dihydrate, unii-52125xyy2a, ccris 6344, disodium chromate dihydrate, sodium dichromate vi dihydrate, sodium dichromate dihydrate vi, chromic acid h2cr2o7 , disodium salt, dihydrate, dichromic acid h2cr2o7 , disodium salt, dihydrate, dsstox_cid_12061, dsstox_rid_78903 | |
JYDRNIYTFCBIFC-UHFFFAOYSA-N | |
disodium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium;dihydrate | |
108005 | |
99.5 to 100.5% | |
Plastic bottle |
357.0°C | |
100 g | |
Cr2Na2O7·2H2O | |
15,8744 | |
Solubility in water: 730g/L (20°C). Other solubilities: insoluble in alcohol | |
O.O.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[Na+].[Na+] | |
298 | |
298 | |
ACS Reagent | |
Sodium dichromate dihydrate |
Safety and Handling
GHS H Statement
May cause genetic defects.
May cause an allergic skin reaction.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Har
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
GHS Signal Word: Danger
RUO – Research Use Only