Please login to your online account to display your discounted pricing

Sodium dichloroacetate, 98%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  338280100

 View more versions of this product

Catalog No. AC338280100

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Powder
Formula Weight 150.92
Quantity 10g
Color White
Beilstein 02, III, 454
EINECS Number 218-461-3
RTECS Number AG9275000
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Chemical Name or Material Sodium dichloroacetate
Infrared Spectrum Authentic
Name Note 98%
Assay Percent Range 98%
Molecular Formula C2HCl2NaO2
Linear Formula NaOC(O)CH(Cl)
MDL Number MFCD00070489
CAS 2156-56-1
Synonym DCA
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Health Hazard 1 HYGROSCOPIC

Specifications

Chemical Name or Material Sodium dichloroacetate
Name Note 98%
CAS 2156-56-1
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C2HCl2NaO2
Linear Formula NaOC(O)CH(Cl)
Formula Weight 150.92
MDL Number MFCD00070489
Synonym DCA
Accessories

HYGROSCOPIC

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents