Please login to your online account to display your discounted pricing

Sodium deuteroxide, for NMR, 40 wt.% solution in D2O, 99+% atom D, ACROS Organics™

$128.18 - $128.18

Chemical Identifiers

Chemical Name or Material Sodium deuteroxide
Name Note 99+ atom % D
CAS 14014-06-3
CAS 7789-20-0
Purity Grade For NMR
Molecular Formula DNaO
Linear Formula NaOD
Formula Weight 41
MDL Number MFCD00037669
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC175470500

  SDS
thermo fisher scientific
175470500
Plastic bottle 50g Each for $128.18
Description & Specifications

Specifications

Physical Form Liquid
Formula Weight 41
Purity Grade For NMR
Quantity 50g
Color Clear colorless
EINECS Number 237-825-2
Density 1.4600g/mL
Packaging Plastic bottle
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Danger
MDL Number MFCD00037669
Deuterium (D) 99.0% min.
CAS 7789-20-0
Linear Formula NaOD
Name Note 99+ atom % D
Chemical Name or Material Sodium deuteroxide
CAS 14014-06-3
Molecular Formula DNaO

For NMR

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.