missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Decyl Sulfate, TCI America™

Manufacturer:  TCI America S09755G

 View more versions of this product

Catalog No. S09755G



Specifications

Specifications

Sodium Decyl Sulfate
192°C
142-87-0
MFCD00041881
XZTJQQLJJCXOLP-UHFFFAOYSA-M
sodium;decyl sulfate
23665771
Crystal-Powder at 20°C
White
5g
C10H21NaO4S
sodium decyl sulfate, sodium n-decyl sulfate, decyl sodium sulfate, sulfuric acid, monodecyl ester, sodium salt, sodium decyl sulphate, unii-al92m833sy, n-decyl sodium sulfate, sodium n-decylsulphate, sulfuric acid, monodecyl ester, sodium salt 1:1, sulfuric acid, decyl ester, sodium salt
CCCCCCCCCCOS(=O)(=O)[O-].[Na+]
260.324
260.32
≥98.0% (GC,T)

Chemical Identifiers

142-87-0
260.324
XZTJQQLJJCXOLP-UHFFFAOYSA-M
23665771
CCCCCCCCCCOS(=O)(=O)[O-].[Na+]
C10H21NaO4S
MFCD00041881
sodium decyl sulfate, sodium n-decyl sulfate, decyl sodium sulfate, sulfuric acid, monodecyl ester, sodium salt, sodium decyl sulphate, unii-al92m833sy, n-decyl sodium sulfate, sodium n-decylsulphate, sulfuric acid, monodecyl ester, sodium salt 1:1, sulfuric acid, decyl ester, sodium salt
sodium;decyl sulfate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1679

RTECSNumber : WS7625000

TSCA : Yes

Documents