Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium cyclohexanebutyrate, AAS, Certified Na Content

$28.01 - $28.01

Chemical Identifiers

Chemical Name or Material Sodium cyclohexanebutyrate
CAS 61886-29-1
Name Note Certified Na Content
Purity Grade AAS Grade
Molecular Formula C10H17NaO2
Formula Weight 192.23
MDL Number MFCD00036395
Synonym Cyclohexanebutyric acid sodium salt; Sodium 4-cyclohexylbutyrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA8726902

  SDS
alfa aesar™
8726902
0.5g Each for $28.01
Description & Specifications

Specifications

Formula Weight 192.23
Name Note Certified Na Content
Molecular Formula C10H17NaO2
MDL Number MFCD00036395
Purity Grade AAS Grade
Chemical Name or Material Sodium cyclohexanebutyrate
CAS 61886-29-1
Melting Point 264°C (dec.)
TSCA TSCA
Physical Form Powder
Quantity 0.5g
Synonym Cyclohexanebutyric acid sodium salt; Sodium 4-cyclohexylbutyrate