missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium cyclohexanebutyrate, AAS, Certified Na Content

Manufacturer:  Alfa Aesar™ 8726904

 View more versions of this product

Catalog No. AA8726904



Specifications

Specifications

Sodium cyclohexanebutyrate
61886-29-1
C10H17NaO2
Cyclohexanebutyric acid sodium salt; Sodium 4-cyclohexylbutyrate
C1CCC(CC1)CCCC(=O)[O-].[Na+]
192.234
192.23
264°C (decomposition)
Certified Na Content
AAS Grade
MFCD00036395
GBEFSJZAXXWKIO-UHFFFAOYSA-M
sodium;4-cyclohexylbutanoate
4072353
Powder
2g

Chemical Identifiers

61886-29-1
192.234
GBEFSJZAXXWKIO-UHFFFAOYSA-M
4072353
C1CCC(CC1)CCCC(=O)[O-].[Na+]
C10H17NaO2
MFCD00036395
Cyclohexanebutyric acid sodium salt; Sodium 4-cyclohexylbutyrate
sodium;4-cyclohexylbutanoate
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents