Please login to your online account to display your discounted pricing

Sodium cyanoborohydride, 1M solution in THF, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle in Tin Can

Manufacturer: thermo fisher scientific  AC372451000

 View more versions of this product

Catalog No. AC372451000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 62.84
Quantity 100mL
Merck Index 15, 8742
Packaging AcroSeal™ Glass Bottle in Tin Can
Density 0.9150g/mL
Molecular Formula CH3BNNa
Linear Formula NaBH3CN
CAS 109-99-9
Appearance of Solution Clear colorless solution
Chemical Name or Material Sodium cyanoborohydride
Name Note 1M solution in THF
CAS 25895-60-7
Health Hazard 1 Danger
MDL Number MFCD00003516
Health Hazard 2 GHS P Statement Fatal if inhaled. Fatal in contact with skin. Toxic if swallowed. Causes severe skin burns and eye damage. May cause respiratory irritation. Toxic to aquatic life with long lasting effects. Highly flam
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN: Gently wash with plenty of soap and water. IF INHALED: Remove v

Specifications

Chemical Name or Material Sodium cyanoborohydride
Name Note 1M solution in THF
CAS 25895-60-7
CAS 109-99-9
Molecular Formula CH3BNNa
Linear Formula NaBH3CN
Formula Weight 62.84
MDL Number MFCD00003516
Accessories

Danger

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN: Gently wash with plenty of soap and water. IF INHALED: Remove v

GHS P Statement Fatal if inhaled. Fatal in contact with skin. Toxic if swallowed. Causes severe skin burns and eye damage. May cause respiratory irritation. Toxic to aquatic life with long lasting effects. Highly flam

SDS & Safety Documents