Please login to your online account to display your discounted pricing

Sodium Cyanide (Granular/Technical), Fisher Chemical


Quantity 250g
Packaging Glass Bottle

Manufacturer: fisher scientific company  S283I250

 View more versions of this product

Catalog No. S283I-250

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Sodium Cyanide
CAS 143-33-9
ChemAlert Storage Symbol Blue
Additional Information Vapor Pressure: 1hPa at 817°C
DOT Information DOT Class 6.1, : Poison
Purity Grade Technical Grade
Physical Form Solid
Molecular Formula CNNa
Formula Weight 49.01
MDL Number MFCD00003523
Density 1.6000g/cm³
Identification Pass Test
Packaging Glass Bottle
Vapor Pressure 1 hPa at 817°C
Boiling Point 1497°C
Color White
Melting Point 562°C
pH 11 to 12
Quantity 250g

Specifications

Chemical Name or Material Sodium Cyanide
CAS 143-33-9
Purity Grade Technical Grade
Physical Form Solid
Molecular Formula CNNa
Formula Weight 49.01
MDL Number MFCD00003523
Accessories
 

SDS & Safety Documents