Please login to your online account to display your discounted pricing

Sodium Cyanide (Granular/Technical), Fisher Chemical

$566.40 - $807.00

Chemical Identifiers

Chemical Name or Material Sodium Cyanide
CAS 143-33-9
Purity Grade Technical Grade
Physical Form Solid
Molecular Formula CNNa
Formula Weight 49.01
MDL Number MFCD00003523
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

S283I-250

  SDS
fisher scientific company
S283I250
Glass Bottle 250g Each for $566.40
Case of 6 Each for $3,398.40

S283I-500

  SDS
thermo fisher scientific
FLS283I-500
Glass Bottle 500g Each for $807.00
Description & Specifications

Specifications

Formula Weight 49.01
Melting Point 562°C
Purity Grade Technical Grade
Boiling Point 1497°C
Quantity 250g
DOT Information DOT Class 6.1, : Poison
Physical Form Solid
Molecular Formula CNNa
Color White
pH 11 to 12
Density 1.6000g/cm³
Vapor Pressure 1 hPa at 817°C
Identification Pass Test
Chemical Name or Material Sodium Cyanide
Additional Information Vapor Pressure: 1hPa at 817°C
CAS 143-33-9
MDL Number MFCD00003523
Packaging Glass Bottle
ChemAlert Storage Symbol Blue
Safety and Handling