Please login to your online account to display your discounted pricing

Sodium Cyanide, Granular, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$387.18 - $695.57

Chemical Identifiers

Chemical Name or Material Sodium Cyanide
CAS 143-33-9
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula NaCN
Formula Weight 49.01
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004071

 
avantor performance
366205
Poly Bottle 2kg Each for $695.57

02004072

 
avantor performance
366201
Amber Glass Bottle 500g Each for $387.18
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 49.01
Chemical Name or Material Sodium Cyanide
CAS 143-33-9
Molecular Formula NaCN
Physical Form Granular
Quantity 2kg
Specific Gravity 1L = 1.60kg
Packaging Poly Bottle