Please login to your online account to display your discounted pricing

Sodium Cyanide, Granular, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 500g
Packaging Amber Glass Bottle

Manufacturer: avantor performance  366201

 View more versions of this product

Catalog No. 02004072

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 49.01
Chemical Name or Material Sodium Cyanide
Molecular Formula NaCN
CAS 143-33-9
Packaging Amber Glass Bottle
Physical Form Granular
Specific Gravity 1L = 1.60kg
Quantity 500g

Specifications

Chemical Name or Material Sodium Cyanide
CAS 143-33-9
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula NaCN
Formula Weight 49.01