Please login to your online account to display your discounted pricing

Sodium Cyanide, For Calcium, Magnesium, 2.5% (w/v), LabChem™

$45.77 - $45.77

Chemical Identifiers

Chemical Name or Material Sodium Cyanide
CAS 7732-18-5
CAS Max % 97.1
CAS 143-33-9
CAS Max % 2.5
CAS 1310-73-2
CAS Max % 0.4
Name Note For Calcium, Magnesium
Linear Formula NaCN
Formula Weight 49
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC237007

 
lab chem
LC237007
125mL Each for $45.77
Description & Specifications

Specifications

Formula Weight 49
Chemical Name or Material Sodium Cyanide
CAS Max % 0.4
Decomposition Information Hydrogen cyanide; Nitrogen oxides
Color Clear, colorless
Quantity 125mL
Name Note For Calcium, Magnesium
Physical Form Liquid
Assay 2.5% (w/v)
Linear Formula NaCN
Density 1g/mL
CAS 7732-18-5
UN Number UN3414
Health Hazard 1 Danger
CAS Max % 97.1
Health Hazard 2 GHS H Statement
Toxic if swallowed or in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Harmful to aquatic life.
Identification Passes Test
CAS 143-33-9
Health Hazard 3 GHS P Statement
Avoid breathing mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Immediately call a poison center or doctor.
If on skin: Wash with plenty of soap and water.
Remove/Take off immediately all contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Packaging Poly Bottle
CAS Max % 2.5
Solubility Soluble in water
CAS 1310-73-2
Specific Gravity 1g/mL
Storage Information Room Temperature
Safety and Handling

Danger

GHS P Statement
Avoid breathing mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Immediately call a poison center or doctor.
If on skin: Wash with plenty of soap and water.
Remove/Take off immediately all contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Toxic if swallowed or in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Harmful to aquatic life.