missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium cyanate, 95%

Manufacturer:  Alfa Aesar™ A124130E

 View more versions of this product

Catalog No. AAA124130E



Description

Description

Applications
Sodium cyanate is an ideal nucleophile, and these nucleophilic properties make it a major contributor to the stereospecificity in certain reactions such as in the production of chiral oxazolidone. Sodium cyanate is a useful reagent in producing asymmetrical urea that have a range of biological activity mostly in aryl isocyanate intermediates. Such intermediates as well as sodium cyanate have been used in medicine as a means of counterbalancing carcinogenic effects on the body.

Solubility
Soluble in water 11.6 g/100 mL (25°C). Slightly soluble in ammonia, benzene. Insoluble in diethyl ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

Sodium cyanate
95%
MFCD00011121
14,8604
C(#N)[O-].[Na+]
65.007
CHEBI:38906
550°C
917-61-3
CNNaO
3594963
ZVCDLGYNFYZZOK-UHFFFAOYSA-M
sodium;cyanate
517096
65.01
2500g

Chemical Identifiers

917-61-3
65.007
ZVCDLGYNFYZZOK-UHFFFAOYSA-M
CHEBI:38906
C(#N)[O-].[Na+]
CNNaO
MFCD00011121
517096
sodium;cyanate
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : GS7000000

TSCA : TSCA

SDS Documents