Please login to your online account to display your discounted pricing

Sodium Cobaltinitrite, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$219.24 - $651.06

Chemical Identifiers

Chemical Name or Material Sodium Cobaltinitrite
CAS 13600-98-1
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula Na3Co(NO2)6
Formula Weight 403.94
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004069

 
avantor performance
365604
Poly Bottle 125g Each for $219.24

02004070

 
avantor performance
365601
Poly Bottle 500g Each for $651.06
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 403.94
Chemical Name or Material Sodium Cobaltinitrite
CAS 13600-98-1
Physical Form Powder
Packaging Poly Bottle
Quantity 125g
Molecular Formula Na3Co(NO2)6

    For Determination of Potassium