Please login to your online account to display your discounted pricing

Sodium Cobaltinitrite, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 500g
Packaging Poly Bottle

Manufacturer: avantor performance  365601

 View more versions of this product

Catalog No. 02004070

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 403.94
Chemical Name or Material Sodium Cobaltinitrite
CAS 13600-98-1
Packaging Poly Bottle
Physical Form Powder
Molecular Formula Na3Co(NO2)6
Quantity 500g

Specifications

Chemical Name or Material Sodium Cobaltinitrite
CAS 13600-98-1
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula Na3Co(NO2)6
Formula Weight 403.94

    For Determination of Potassium