missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Citrate USP GenAR Crystal, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 6132-04-3
Physical Form Crystalline
Molecular Formula C6H5Na3O7·2H2O
Formula Weight 294.1
Chemical Name or Material Sodium Citrate, Dihydrate
Grade USP
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK7773500 Avantor Performance
777304
Glass 500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

6132-04-3
C6H5Na3O7·2H2O
Sodium Citrate, Dihydrate
Crystalline
294.1
USP

Specifications

294.1
C6H5Na3O7·2H2O
6132-04-3
Sodium Citrate, Dihydrate
Crystalline
Glass
500g
USP