missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Citrate, For Copper, Certified, 30% (w/v), LabChem™

$224.86 - $224.86

Chemical Identifiers

CAS 7732-18-5,6132-04-3
CAS Max % 74.84,25.16
Linear Formula Na3C6O7H5·2H2O
Formula Weight 294.1
Chemical Name or Material Sodium Citrate, For Copper
Grade Certified Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC236602 Lab Chem
LC236602
1L Each for $224.86
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,6132-04-3
Na3C6O7H5·2H2O
Sodium Citrate, For Copper
74.84,25.16
294.1
Certified Grade

Specifications

7732-18-5,6132-04-3
Liquid
294.1
Passes Test
Soluble in water
Passes Test
Fume; Carbon monoxide; Carbon dioxide
1L
30% (w/v)
74.84,25.16
Na3C6O7H5·2H2O
1.2g/mL
Poly Bottle
1.2g/mL
Room Temperature
Clear, colorless
Sodium Citrate, For Copper
Certified Grade

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.