missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Citrate, Dihydrate, Powder, U.S.P. - F.C.C., J.T. Baker™
Supplier: Avantor 365005
Specifications
Sodium Citrate | |
6132-04-3 | |
2.5 kg | |
trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate | |
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+] | |
294.10 | |
CHEBI:32142 | |
FCC/USP | |
1L = 1.70kg |
Dihydrate | |
Powder | |
C6H9Na3O9 | |
NLJMYIDDQXHKNR-UHFFFAOYSA-K | |
trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate | |
71474 | |
294.1 | |
Poly Bottle |
Chemical Identifiers
6132-04-3 | |
294.10 | |
trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate | |
CHEBI:32142 | |
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+] |
C6H9Na3O9 | |
NLJMYIDDQXHKNR-UHFFFAOYSA-K | |
71474 | |
trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate |