missing translation for 'onlineSavingsMsg'
Learn More
missing translation for 'pleaseLogIn'

Sodium Citrate, Dihydrate, Granular, Ultrapure Bioreagent, J.T.Baker™

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

$275.05 - $540.70

Chemical Identifiers

CAS 6132-04-3
Molecular Formula C6H5Na3O7·2H2O
Formula Weight 294.1
Chemical Name or Material Sodium Citrate, Dihydrate
Grade Ultrapure Bioreagent
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Qty  
Catalog Number Mfr. No. Packaging Quantity Price Qty  
02004066 Avantor Performance
409306
Poly Pail 5kg Each for $275.05
02004060 Avantor Performance
409304
Poly Bottle 1kg Each for $540.70
02004065 Avantor Performance
409301
Poly Bottle 500g Each for $409.00
Specifications

Description

    For Liquid Chromatography and Molecular Biology Applications
Description & Specifications

Chemical Identifiers

6132-04-3
294.1
Ultrapure Bioreagent
C6H5Na3O7·2H2O
Sodium Citrate, Dihydrate

Specifications

6132-04-3
C6H5Na3O7·2H2O
Poly Pail
5kg
Ultrapure Bioreagent
Granular
294.1
1L = 1.70kg
Sodium Citrate, Dihydrate