Please login to your online account to display your discounted pricing

Sodium Citrate, Dihydrate, Granular, Ultrapure Bioreagent, J.T.Baker™


High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

Quantity 5kg
Packaging Poly Pail

Manufacturer: avantor performance  409306

 View more versions of this product

Catalog No. 02004066

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Sodium Citrate
Name Note Dihydrate
CAS 6132-04-3
Purity Grade Ultrapure Bioreagent
Physical Form Granular
Molecular Formula C6H5Na3O7·2H2O
Formula Weight 294.1
Packaging Poly Pail
Specific Gravity 1L = 1.70kg
Quantity 5kg

Specifications

Chemical Name or Material Sodium Citrate
CAS 6132-04-3
Name Note Dihydrate
Purity Grade Ultrapure Bioreagent
Molecular Formula C6H5Na3O7·2H2O
Formula Weight 294.1

    For Liquid Chromatography and Molecular Biology Applications