Please login to your online account to display your discounted pricing

Sodium Citrate, Dihydrate, ACS Grade, ≥ 99.0%, LabChem™

$68.26 - $68.26

Chemical Identifiers

Chemical Name or Material Sodium Citrate
CAS 6132-04-3
CAS Max % 100
Name Note Dihydrate
Purity Grade ACS Grade
Linear Formula Na3C6O7H5·2H2O
Formula Weight 294.1
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC236501

 
lab chem
LC236501
500g Each for $68.26
Description & Specifications

Specifications

Assay ≥99.0%
Formula Weight 294.1
Chemical Name or Material Sodium Citrate
Identification Passes Test
Name Note Dihydrate
Packaging Poly Bottle
Purity Grade ACS Grade
Physical Form Crystals
Solubility Soluble in water
Specific Gravity 1.7g/cm3
Storage Information Room Temperature
Linear Formula Na3C6O7H5·2H2O
CAS 6132-04-3
Decomposition Information Fume; Carbon monoxide; Carbon dioxide
Color White
Melting Point 150°C
Quantity 500g
CAS Max % 100
Health Hazard 2 GHS H Statement
Product is not hazardous.
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Density 1.7g/cm3
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Product is not hazardous.