missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Citrate ACS AR Crystal, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 6132-04-3
Physical Form Crystalline
Molecular Formula HOC(COONa)(CH2COONa)2·2H2O
Formula Weight 294.1
Chemical Name or Material Sodium Citrate, Dihydrate
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK0754212 Avantor Performance
075406
Poly 2.5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MK0754500 Avantor Performance
075412
POLYSTORMOR 500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

6132-04-3
HOC(COONa)(CH2COONa)2·2H2O
Sodium Citrate, Dihydrate
Crystalline
294.1
ACS Grade

Specifications

294.1
6132-04-3
HOC(COONa)(CH2COONa)2·2H2O
Sodium Citrate, Dihydrate
2.5kg
Crystalline
Poly
ACS Grade