missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Chromate Anhydrous (Powder/Certified), Fisher Chemical

Manufacturer:  Thermo Scientific S272500

 View more versions of this product

Catalog No. S272-500




Specifications

Specifications

Sodium Chromate Anhydrous
Pass Test
MFCD00003484
YZHUMGUJCQRKBT-UHFFFAOYSA-M
sodium;chlorate
516902
161.97
Certified
0.01% max.
1.0% max.
Yellow
9.2
7775-09-9
ClNaO3
sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal
[O-]Cl(=O)=O.[Na+]
106.44
CHEBI:65242
Solid
0.005% max.
Pass Test
Glass Bottle
792°C
500g

Chemical Identifiers

7775-09-9
106.44
YZHUMGUJCQRKBT-UHFFFAOYSA-M
516902
sodium;chlorate
ClNaO3
MFCD00003484
sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal
CHEBI:65242
[O-]Cl(=O)=O.[Na+]
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 5.1, : Oxidizer

SDS Documents