Please login to your online account to display your discounted pricing

Sodium Chromate Anhydrous (Powder/Certified), Fisher Chemical


Quantity 250g
Packaging Glass Bottle

Manufacturer: fisher scientific company  S272250

 View more versions of this product

Catalog No. S272-250

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified Grade
Formula Weight 161.97
Melting Point 792°C
Quantity 250g
DOT Information DOT Class 5.1, : Oxidizer
Physical Form Solid
Molecular Formula CrNa2O4
Color Yellow
pH 9.2
Identification Pass Test
Loss on Drying 1.0% max.
ChemAlert Storage Symbol Yellow
Synonym Chromic Acid Disodium Salt
Chloride 0.01% max.
Chemical Name or Material Sodium Chromate Anhydrous
Aluminum (Al) Pass Test
Sulfate 0.01% max.
MDL Number MFCD00003484
Calcium (Ca) 0.005% max.
Packaging Glass Bottle
CAS 7775-11-3

Specifications

Chemical Name or Material Sodium Chromate Anhydrous
CAS 7775-11-3
Purity Grade Certified Grade
Physical Form Solid
Molecular Formula CrNa2O4
Formula Weight 161.97
MDL Number MFCD00003484
Synonym Chromic Acid Disodium Salt
Accessories
 

SDS & Safety Documents