Please login to your online account to display your discounted pricing

Sodium Chromate Anhydrous (Powder/Certified), Fisher Chemical

$329.69 - $401.54

Chemical Identifiers

Chemical Name or Material Sodium Chromate Anhydrous
CAS 7775-11-3
Purity Grade Certified Grade
Physical Form Solid
Molecular Formula CrNa2O4
Formula Weight 161.97
MDL Number MFCD00003484
Synonym Chromic Acid Disodium Salt
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

S272-250

  SDS
fisher scientific company
S272250
Glass Bottle 250g Each for $329.69
Case of 6 Each for $1,978.14

S272-500

  SDS
fisher scientific company
S272500
Glass Bottle 500g Each for $401.54
Case of 6 Each for $2,409.24
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 161.97
Melting Point 792°C
Quantity 250g
DOT Information DOT Class 5.1, : Oxidizer
Physical Form Solid
Molecular Formula CrNa2O4
Viscosity [NULL]
Color Yellow
pH 9.2
Identification Pass Test
Loss on Drying 1.0% max.
ChemAlert Storage Symbol Yellow
Synonym Chromic Acid Disodium Salt
Chloride 0.01% max.
Chemical Name or Material Sodium Chromate Anhydrous
Aluminum (Al) Pass Test
Sulfate 0.01% max.
MDL Number MFCD00003484
Calcium (Ca) 0.005% max.
Packaging Glass Bottle
CAS 7775-11-3
Safety and Handling