missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium chromate, anhydrous, 98%

$58.74 - $894.43

Chemical Identifiers

CAS 7775-09-9
Molecular Formula ClNaO3
Molecular Weight (g/mol) 106.44
MDL Number MFCD00003484
InChI Key YZHUMGUJCQRKBT-UHFFFAOYSA-M
Synonym sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal
PubChem CID 516902
ChEBI CHEBI:65242
IUPAC Name sodium;chlorate
SMILES [O-]Cl(=O)=O.[Na+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1054736 View Documents Alfa Aesar™
A1054736
500g Each for $220.53
 
AAA1054722 View Documents Alfa Aesar™
A1054722
100g Each for $58.74
 
AAA105470E View Documents Alfa Aesar™
A105470E
2500g Each for $894.43
 
Description

Description

Applications
Sodium chromate anhydrous is used as a corrosion inhibitor in the petroleum industry, wood preservative and dyeing auxiliary in the textile industry. It is also used to prepare pigments for paints and inks. It also finds use in pharmaceuticals to determine red blood cell volume. In synthetic chemistry, it is utilized as an oxidant to convert primary alcohols and secondary alcohols to carboxylic acids and ketones respectively.

Solubility
Soluble in water. Slightly soluble in methanol and ethanol.

Notes
Hygroscopic. Incompatible with combustible materials and reducing agents.
Specifications

Chemical Identifiers

7775-09-9
106.44
YZHUMGUJCQRKBT-UHFFFAOYSA-M
516902
sodium;chlorate
ClNaO3
MFCD00003484
sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal
CHEBI:65242
[O-]Cl(=O)=O.[Na+]

Specifications

7775-09-9
MFCD00003484
14,8601
YZHUMGUJCQRKBT-UHFFFAOYSA-M
sodium;chlorate
516902
161.97
Odorless
500g
ClNaO3
UN3288
sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal
[O-]Cl(=O)=O.[Na+]
106.44
CHEBI:65242
98%
792°C
Sodium chromate, Anhydrous
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H330-H334-H340-H350-H360-H372-H314-H318-H317
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

GHS P Statement
P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

RTECSNumber : GB2955000

TSCA : TSCA