Please login to your online account to display your discounted pricing

Sodium Chromate, 4-Hydrate, Crystal, BAKER ANALYZED™ Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 125g
Packaging Poly Bottle

Manufacturer: avantor performance  364004

 View more versions of this product

Catalog No. 02004051

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystals
Formula Weight 234.03
Chemical Name or Material Sodium Chromate
Name Note 4-Hydrate
Packaging Poly Bottle
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula Na2CrO4·4H2O
CAS 10034-82-9
Quantity 125g

Specifications

Chemical Name or Material Sodium Chromate
CAS 10034-82-9
Name Note 4-Hydrate
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula Na2CrO4·4H2O
Formula Weight 234.03